Ethyl-2-cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Supplier: Matrix Scientific
080882-1G
080882-5G
080882-500MG
102671-732EA
193.85
USD
102671-732
102671-734
102671-736
Ethyl-2-cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-2-cyclobutyl-7-[(E)-2-(dimethylamino)vinyl][1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Hazard: Irritant
Formula:
C₁₆H₂₁N₅O₂ |
MDL Number:
MFCD22374991 CAS Number: 1379821-55-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...