Ethyl-2-cyclobutyl-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Supplier: Matrix Scientific

080881-500MG 080881-1G 080881-5G
102671-730EA 1130.77 USD
102671-730 102671-726 102671-728
Ethyl-2-cyclobutyl-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate
Ethyl-2-cyclobutyl-7-methyl[1,2,4]triazolo[1,5-a]pyrimidine-6-carboxylate

Hazard: Irritant

Formula: C₁₃H₁₆N₄O₂
MDL Number: MFCD22374990
CAS Number: 1379811-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR