N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)methanesulfonamide
Supplier: Matrix Scientific
080872-500MG
080872-5G
080872-1G
102671-682EA
1615.38
USD
102671-682
102671-678
102671-680
N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)methanesulfonamide
N-(5-Cyclopropyl-1,4,5,6-tetrahydro-1,3,5-triazin-2-yl)methanesulfonamide
Hazard: Irritant
Formula:
C₇H₁₄N₄O₂S |
MDL Number:
MFCD15195698 CAS Number: 1209203-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...