2-(Cyclopropylamino)-6-(methoxymethyl)pyrimidin-4(3H)-one
Supplier: Matrix Scientific
080866-500MG
080866-5G
080866-1G
102671-642EA
258.46
USD
102671-642
102671-644
102671-646
2-(Cyclopropylamino)-6-(methoxymethyl)pyrimidin-4(3H)-one
2-(Cyclopropylamino)-6-(methoxymethyl)pyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₉H₁₃N₃O₂ |
MDL Number:
MFCD20293197 CAS Number: 1158516-14-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...