2-(Cyclopropylamino)-4-methylpyrimidine-5-carboxylic acid
Supplier: Matrix Scientific
080863-500MG
080863-5G
080863-1G
102671-624EA
387.69
USD
102671-624
102671-626
102671-628
2-(Cyclopropylamino)-4-methylpyrimidine-5-carboxylic acid
2-(Cyclopropylamino)-4-methylpyrimidine-5-carboxylic acid
Hazard: Irritant
Formula:
C₉H₁₁N₃O₂ |
MDL Number:
MFCD20293202 CAS Number: 1343961-35-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...