6-Cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylic acid
Supplier: Matrix Scientific
080859-1G
080859-500MG
102671-602EA
387.69
USD
102671-602
102671-604
6-Cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylic acid
6-Cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylic acid
Hazard: Irritant
Formula:
C₁₃H₁₂N₂O₃ |
MDL Number:
MFCD22374978 CAS Number: 1379811-67-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...