Methyl-6-cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080858-1G 080858-5G 080858-500MG
102671-596EA 258.46 USD
102671-596 102671-598 102671-600
Methyl-6-cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-6-cyclopropyl-2-methyl-5-oxo-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₄H₁₄N₂O₃
MDL Number: MFCD13872541
CAS Number: 1158282-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR