N-[4-(1,3-Benzodioxol-5-yl)-1,3-thiazol-2-yl]guanidine
Supplier: Matrix Scientific
080851-1G
080851-500MG
102671-558EA
387.69
USD
102671-558
102671-560
N-[4-(1,3-Benzodioxol-5-yl)-1,3-thiazol-2-yl]guanidine
N-[4-(1,3-Benzodioxol-5-yl)-1,3-thiazol-2-yl]guanidine
Hazard: Irritant
Formula:
C₁₁H₁₀N₄O₂S |
MDL Number:
MFCD06739733 CAS Number: 886497-71-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...