N-[2-(2-Methoxyphenyl)ethyl]guanidine sulfate
Supplier: Matrix Scientific
080849-5G
080849-500MG
080849-1G
102671-550EA
323.08
USD
102671-550
102671-552
102671-548
N-[2-(2-Methoxyphenyl)ethyl]guanidine sulfate
N-[2-(2-Methoxyphenyl)ethyl]guanidine sulfate
Hazard: Irritant
Formula:
C₁₀H₁₇N₃O₅S |
MDL Number:
MFCD22422008 CAS Number: 1417567-55-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...