N-(6-Bromo-4-hydroxyquinazolin-2-yl)guanidine

Supplier: Matrix Scientific

080846-500MG 080846-1G
102671-534EA 258.46 USD
102671-534 102671-536
N-(6-Bromo-4-hydroxyquinazolin-2-yl)guanidine
N-(6-Bromo-4-hydroxyquinazolin-2-yl)guanidine

Hazard: Irritant

Formula: C₉H₈BrN₅O
MDL Number: MFCD05724784
CAS Number: 74189-10-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR