N-[4-(4-Methoxyphenyl)-1,3-thiazol-2-yl]guanidine
Supplier: Matrix Scientific
080843-5G
080843-500MG
080843-1G
102671-520EA
323.08
USD
102671-520
102671-522
102671-518
N-[4-(4-Methoxyphenyl)-1,3-thiazol-2-yl]guanidine
N-[4-(4-Methoxyphenyl)-1,3-thiazol-2-yl]guanidine
Hazard: Irritant
Formula:
C₁₁H₁₂N₄OS |
MDL Number:
MFCD06739725 CAS Number: 91089-11-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...