N-[4-(4-Methylphenyl)-1,3-thiazol-2-yl]guanidine
Supplier: Matrix Scientific
080840-1G
080840-500MG
102671-502EA
193.85
USD
102671-502
102671-504
N-[4-(4-Methylphenyl)-1,3-thiazol-2-yl]guanidine
N-[4-(4-Methylphenyl)-1,3-thiazol-2-yl]guanidine
Hazard: Irritant
Formula:
C₁₁H₁₂N₄S |
MDL Number:
MFCD00662566 CAS Number: 7120-04-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...