4-Pyridin-2-ylpiperazine-1-carboximidamide sulfate
Supplier: Matrix Scientific
080836-1G
080836-500MG
102671-486EA
193.85
USD
102671-486
102671-488
4-Pyridin-2-ylpiperazine-1-carboximidamide sulfate
4-Pyridin-2-ylpiperazine-1-carboximidamide sulfate
Hazard: Irritant
Formula:
C₁₀H₁₇N₅O₄S |
MDL Number:
MFCD22422003 CAS Number: 1187455-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...