Methyl [(4Z)-4-[(dimethylamino)methylene]-1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate

Supplier: Matrix Scientific

080793-500MG
102671-252EA 258.46 USD
102671-252
Methyl [(4Z)-4-[(dimethylamino)methylene]-1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate
Methyl [(4Z)-4-[(dimethylamino)methylene]-1-(4-methoxyphenyl)-5-oxo-4,5-dihydro-1H-pyrazol-3-yl]acetate

Hazard: Irritant

Formula: C₁₆H₁₉N₃O₄
MDL Number: MFCD22374961
CAS Number: 1379821-56-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR