Tetramethyl-5',5',8'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Supplier: Matrix Scientific
080781-500MG
080781-1G
102671-214EA
161.54
USD
102671-214
102671-216
Tetramethyl-5',5',8'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Tetramethyl-5',5',8'-trimethyl-5',6'-dihydrospiro[1,3-dithiole-2,1'-thiopyrano[2,3-c]quinoline]-2',3',4,5-tetracarboxylate
Hazard: Irritant
Formula:
C₂₅H₂₅NO₈S₃ MW: 563.67 g/mol |
MDL Number:
MFCD00532515 CAS Number: 305343-76-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...