6-(Chloromethyl)-N-(4-phenoxyphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080737-500MG
102671-020EA 258.46 USD
102671-020
6-(Chloromethyl)-N-(4-phenoxyphenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(4-phenoxyphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₆H₁₄ClN₅O
MDL Number: MFCD03941307
CAS Number: 644959-89-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR