6-(Chloromethyl)-N-(2-ethoxyphenyl)-1,3,5-triazine-2,4-diamine

Supplier: Matrix Scientific

080725-500MG
102670-972EA 258.46 USD
102670-972
6-(Chloromethyl)-N-(2-ethoxyphenyl)-1,3,5-triazine-2,4-diamine
6-(Chloromethyl)-N-(2-ethoxyphenyl)-1,3,5-triazine-2,4-diamine

Hazard: Irritant

Formula: C₁₂H₁₄ClN₅O
MDL Number: MFCD22374947
CAS Number: 1379811-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR