4-(1,3-Benzodioxol-5-yl)-2,5-dimethyl-3-furoic acid

Supplier: Matrix Scientific

080646-500MG 080646-1G
102670-558EA 516.92 USD
102670-558 102670-560
4-(1,3-Benzodioxol-5-yl)-2,5-dimethyl-3-furoic acid
4-(1,3-Benzodioxol-5-yl)-2,5-dimethyl-3-furoic acid

Hazard: Irritant

Formula: C₁₄H₁₂O₅
MDL Number: MFCD20129826
CAS Number: 104794-31-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR