2-Imino-7,7-dimethyl-4-phenyl-2,6,7,8-tetrahydro-5H-1,3-benzothiazin-5-one
Supplier: Matrix Scientific
080618-1G
080618-500MG
102670-420EA
243.08
USD
102670-420
102670-418
2-Imino-7,7-dimethyl-4-phenyl-2,6,7,8-tetrahydro-5H-1,3-benzothiazin-5-one
2-Imino-7,7-dimethyl-4-phenyl-2,6,7,8-tetrahydro-5H-1,3-benzothiazin-5-one
Hazard: Irritant
Formula:
C₁₆H₁₆N₂OS |
MDL Number:
MFCD22374926 CAS Number: 1374509-65-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...