1-Acetyl-3-phenyl-1,5,6,7-tetrahydro-4H-indazol-4-one

Supplier: Matrix Scientific

080614-1G 080614-500MG
102670-404EA 323.08 USD
102670-404 102670-402
1-Acetyl-3-phenyl-1,5,6,7-tetrahydro-4H-indazol-4-one
1-Acetyl-3-phenyl-1,5,6,7-tetrahydro-4H-indazol-4-one

Hazard: Irritant

Formula: C₁₅H₁₄N₂O₂
MDL Number: MFCD22374922
CAS Number: 1374509-31-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR