Methyl-1-benzyl-7-methyl-2,4-dioxo-1,2,3,4-tetrahydro-pyrido[2,3-d]pyrimidine-6-carboxylate
Supplier: Matrix Scientific
080589-500MG
102670-292EA
193.85
USD
102670-292
Methyl-1-benzyl-7-methyl-2,4-dioxo-1,2,3,4-tetrahydro-pyrido[2,3-d]pyrimidine-6-carboxylate
Methyl-1-benzyl-7-methyl-2,4-dioxo-1,2,3,4-tetrahydro-pyrido[2,3-d]pyrimidine-6-carboxylate
Hazard: Irritant
Formula:
C₁₇H₁₅N₃O₄ |
MDL Number:
MFCD22374915 CAS Number: 1374509-50-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...