1-(2-Furoyl)-2-methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine
Supplier: Matrix Scientific
080579-1G
080579-500MG
102670-256EA
161.54
USD
102670-256
102670-258
1-(2-Furoyl)-2-methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine
1-(2-Furoyl)-2-methyl-N-phenyl-1,2,3,4-tetrahydroquinolin-4-amine
Hazard: Irritant
Formula:
C₂₁H₂₀N₂O₂ |
MDL Number:
MFCD00627087 CAS Number: 301655-67-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...