5-Butyl-4-chloro-6-methylpyrimidin-2-amine

Supplier: Matrix Scientific

080527-500MG
102669-982EA 387.69 USD
102669-982
5-Butyl-4-chloro-6-methylpyrimidin-2-amine
5-Butyl-4-chloro-6-methylpyrimidin-2-amine

Hazard: Irritant

Formula: C₉H₁₄ClN₃
MDL Number: MFCD00460874
CAS Number: 162272-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR