2-[(4,7-Dimethylquinazolin-2-yl)amino]-6-methylpyrimidin-4(3H)-one
Supplier: Matrix Scientific
080517-1G
080517-500MG
102669-952EA
193.85
USD
102669-952
102669-954
2-[(4,7-Dimethylquinazolin-2-yl)amino]-6-methylpyrimidin-4(3H)-one
2-[(4,7-Dimethylquinazolin-2-yl)amino]-6-methylpyrimidin-4(3H)-one
Hazard: Irritant
Formula:
C₁₅H₁₅N₅O |
MDL Number:
MFCD22374899 CAS Number: 332074-12-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...