1-[7-Methyl-2-(4-methylphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone

Supplier: Matrix Scientific

080502-500MG 080502-1G
102669-902EA 258.46 USD
102669-902 102669-904
1-[7-Methyl-2-(4-methylphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone
1-[7-Methyl-2-(4-methylphenyl)[1,2,4]triazolo[1,5-a]pyrimidin-6-yl]ethanone

Hazard: Irritant

Formula: C₁₅H₁₄N₄O
MDL Number: MFCD07469694
CAS Number: 886152-33-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR