Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-(4H-1,2,4-triazol-4-yl)-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Supplier: Matrix Scientific

080479-500MG 080479-1G
102669-818EA 243.08 USD
102669-818 102669-816
Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-(4H-1,2,4-triazol-4-yl)-5,6-dihydro-1,6-naphthyridine-3-carboxylate
Methyl-2-[(E)-2-(dimethylamino)vinyl]-5-oxo-6-(4H-1,2,4-triazol-4-yl)-5,6-dihydro-1,6-naphthyridine-3-carboxylate

Hazard: Irritant

Formula: C₁₆H₁₆N₆O₃
MDL Number: MFCD22374881
CAS Number: 1374510-79-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR