2-(3,4-Dihydroisoquinolin-2(1H)-yl)-4-methylpyrimidine-5-carboxylic acid
Supplier: Matrix Scientific
080462-500MG
080462-1G
102669-762EA
323.08
USD
102669-762
102669-760
2-(3,4-Dihydroisoquinolin-2(1H)-yl)-4-methylpyrimidine-5-carboxylic acid
2-(3,4-Dihydroisoquinolin-2(1H)-yl)-4-methylpyrimidine-5-carboxylic acid
Hazard: Irritant
Formula:
C₁₅H₁₅N₃O₂ |
MDL Number:
MFCD10006001 CAS Number: 1017485-42-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...