(2Z,2'Z)-2,2'-(6-Nitro-1,4-dihydroquinoxaline-2,3-diylidene)bis(1-phenylethanone)

Supplier: Matrix Scientific

080435-1G 080435-5G 080435-500MG
102669-652EA 243.08 USD
102669-652 102669-654 102669-650
(2Z,2'Z)-2,2'-(6-Nitro-1,4-dihydroquinoxaline-2,3-diylidene)bis(1-phenylethanone)
(2Z,2'Z)-2,2'-(6-Nitro-1,4-dihydroquinoxaline-2,3-diylidene)bis(1-phenylethanone)

Hazard: Irritant

Formula: C₂₄H₁₇N₃O₄
MDL Number: MFCD22374860
CAS Number: 150012-79-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR