[1-Cyclopropyl-2-(4-methylphenoxy)ethyl]amine

Supplier: Matrix Scientific

080424-1G 080424-500MG
102669-590EA 516.92 USD
102669-590 102669-592
[1-Cyclopropyl-2-(4-methylphenoxy)ethyl]amine
[1-Cyclopropyl-2-(4-methylphenoxy)ethyl]amine

Hazard: Irritant

Formula: C₁₂H₁₇NO
MDL Number: MFCD19605955
CAS Number: 1342559-22-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR