[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]methylamine
Supplier: Matrix Scientific
080416-500MG
080416-1G
102669-554EA
484.62
USD
102669-554
102669-552
[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]methylamine
[1-Cyclopropyl-2-(2-methylphenoxy)ethyl]methylamine
Hazard: Irritant
Formula:
C₁₃H₁₉NO |
MDL Number:
MFCD19605924 CAS Number: 1339577-19-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...