3-[(5-Methyl-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Supplier: Matrix Scientific

080372-1G 080372-500MG
102669-324EA 387.69 USD
102669-324 102669-326
3-[(5-Methyl-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
3-[(5-Methyl-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine

Hazard: Irritant

Formula: C₁₂H₁₇N₅
MDL Number: MFCD12405271
CAS Number: 1174847-42-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR