3-[(3-Methyl-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
Supplier: Matrix Scientific
080363-500MG
080363-1G
102669-288EA
387.69
USD
102669-288
102669-290
3-[(3-Methyl-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
3-[(3-Methyl-4-nitro-1H-pyrazol-1-yl)methyl]-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
Hazard: Irritant
Formula:
C₁₂H₁₆N₆O₂ |
MDL Number:
MFCD16345370 CAS Number: 1245772-82-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...