3-(1-Ethyl-4-nitro-1H-pyrazol-3-yl)-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
Supplier: Matrix Scientific
080361-1G
080361-500MG
080361-5G
102669-278EA
387.69
USD
102669-278
102669-282
102669-280
3-(1-Ethyl-4-nitro-1H-pyrazol-3-yl)-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
3-(1-Ethyl-4-nitro-1H-pyrazol-3-yl)-6,7,8,9-tetrahydro-5H-[1,2,4]triazolo[4,3-a]azepine
Hazard: Irritant
Formula:
C₁₂H₁₆N₆O₂ |
MDL Number:
MFCD16345367 CAS Number: 1245772-81-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...