Ethyl-2-(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)propanoate
Supplier: Matrix Scientific
080341-500MG
080341-1G
080341-5G
102669-194EA
646.15
USD
102669-194
102669-196
102669-192
Ethyl-2-(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)propanoate
Ethyl-2-(3,5-dimethyl-4-nitro-1H-pyrazol-1-yl)propanoate
Hazard: Irritant
Formula:
C₁₀H₁₅N₃O₄ |
MDL Number:
MFCD00206698 CAS Number: 494747-21-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...