(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
Supplier: Matrix Scientific
080327-500MG
080327-5G
080327-1G
102669-120EA
646.15
USD
102669-120
102669-122
102669-118
(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
(2-{[1-Ethyl-3-(trifluoromethyl)-1H-pyrazol-5-yl]methoxy}ethyl)amine hydrochloride
Hazard: Irritant
Formula:
C₉H₁₅ClF₃N₃O MW: 273.69 g/mol |
MDL Number:
MFCD22421975 CAS Number: 1417570-03-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...