4-Cyclopropyl-6-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

Supplier: Matrix Scientific

080283-1G
102668-856EA 484.62 USD
102668-856
4-Cyclopropyl-6-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine
4-Cyclopropyl-6-(trifluoromethyl)-1H-pyrazolo[3,4-b]pyridin-3-amine

Hazard: Irritant

Formula: C₁₀H₉F₃N₄
MDL Number: MFCD04967270
CAS Number: 832739-88-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR