(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride

Supplier: Matrix Scientific

Synonyms: 2-(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride

080268-500MG 080268-5G
102668-764EA 338.46 USD
102668-764 102668-768
(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride
(4-Amino-1H-pyrazol-1-yl)acetic acid hydrochloride

Hazard: Irritant

Formula: C₅H₈ClN₃O₂
MW: 177.59 g/mol
MDL Number: MFCD22421967
CAS Number: 1417569-73-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR