1-(Carboxymethyl)-3-nitro-1H-pyrazole-5-carboxylic acid

Supplier: Matrix Scientific

080261-1G
102668-724EA 646.15 USD
102668-724
1-(Carboxymethyl)-3-nitro-1H-pyrazole-5-carboxylic acid
1-(Carboxymethyl)-3-nitro-1H-pyrazole-5-carboxylic acid

Hazard: Irritant

Formula: C₆H₅N₃O₆
MDL Number: MFCD06739328
CAS Number: 956439-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR