N-[(3,5-Dimethyl-1H-pyrazol-1-yl)(imino)methyl]benzamide
Supplier: Matrix Scientific
080239-1G
102668-598EA
276.92
USD
102668-598
N-[(3,5-Dimethyl-1H-pyrazol-1-yl)(imino)methyl]benzamide
N-[(3,5-Dimethyl-1H-pyrazol-1-yl)(imino)methyl]benzamide
Hazard: Irritant
Formula:
C₁₃H₁₄N₄O |
MDL Number:
MFCD22374851 CAS Number: 51883-88-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...