[(5-Isopropyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Supplier: Matrix Scientific

080223-1G
102668-508EA 484.62 USD
102668-508
[(5-Isopropyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid
[(5-Isopropyl-1,3,4-thiadiazol-2-yl)amino](oxo)acetic acid

Hazard: Irritant

Formula: C₇H₉N₃O₃S
MDL Number: MFCD15181807
CAS Number: 1158206-46-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR