Ethyl oxo{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]amino}acetate

Supplier: Matrix Scientific

080218-1G
102668-484EA 484.62 USD
102668-484
Ethyl oxo{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]amino}acetate
Ethyl oxo{[5-(trifluoromethyl)-1,3,4-thiadiazol-2-yl]amino}acetate

Hazard: Irritant

Formula: C₇H₆F₃N₃O₃S
MDL Number: MFCD00111799
CAS Number: 519170-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR