Ethyl oxo[(2-oxotetrahydro-3-thienyl)amino]acetate

Supplier: Matrix Scientific

080217-1G
102668-480EA 484.62 USD
102668-480
Ethyl oxo[(2-oxotetrahydro-3-thienyl)amino]acetate
Ethyl oxo[(2-oxotetrahydro-3-thienyl)amino]acetate

Hazard: Irritant

Formula: C₈H₁₁NO₄S
MDL Number: MFCD02625497
CAS Number: 956361-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR