N-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxamide

Supplier: Matrix Scientific

077919-1G
102668-334EA 323.08 USD
102668-334
N-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxamide
N-(3,4-Dichlorophenyl)-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₁₀H₇Cl₂N₃O
MDL Number: MFCD02128535
CAS Number: 305346-22-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR