N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-1H-pyrazole-3-carboxamide

Supplier: Matrix Scientific

077911-1G
102668-298EA 243.08 USD
102668-298
N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-1H-pyrazole-3-carboxamide
N-(1,5-Dimethyl-3-oxo-2-phenyl-2,3-dihydro-1H-pyrazol-4-yl)-1H-pyrazole-3-carboxamide

Hazard: Irritant

Formula: C₁₅H₁₅N₅O₂
MDL Number: MFCD02608726
CAS Number: 379255-85-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR