N~1~-(4-Methoxybenzyl)-4-(trifluoromethyl)benzene-1,2-diamine

Supplier: Matrix Scientific

077893-500MG
102668-226EA 193.85 USD
102668-226
N~1~-(4-Methoxybenzyl)-4-(trifluoromethyl)benzene-1,2-diamine
N~1~-(4-Methoxybenzyl)-4-(trifluoromethyl)benzene-1,2-diamine

Hazard: Irritant

Formula: C₁₅H₁₅F₃N₂O
MDL Number: MFCD13379217
CAS Number: 780010-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR