2-(2-Hydroxypropyl)-1H-isoindole-1,3(2H)-dione

Supplier: Matrix Scientific

077888-500MG 077888-1G
102668-204EA 516.92 USD
102668-204 102668-206
2-(2-Hydroxypropyl)-1H-isoindole-1,3(2H)-dione
2-(2-Hydroxypropyl)-1H-isoindole-1,3(2H)-dione

Hazard: Irritant

Formula: C₁₁H₁₁NO₃
MDL Number: MFCD14587522
CAS Number: 3700-55-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR