Ethyl [(1-ethylpropyl)amino](oxo)acetate

Supplier: Matrix Scientific

077867-1G
102668-112EA 484.62 USD
102668-112
Ethyl [(1-ethylpropyl)amino](oxo)acetate
Ethyl [(1-ethylpropyl)amino](oxo)acetate

Hazard: Irritant

Formula: C₉H₁₇NO₃
MDL Number: MFCD02862419
CAS Number: 349442-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR