Ethyl (isobutylamino)(oxo)acetate

Supplier: Matrix Scientific

077863-1G
102668-090EA 484.62 USD
102668-090
Ethyl (isobutylamino)(oxo)acetate
Ethyl (isobutylamino)(oxo)acetate

Hazard: Irritant

Formula: C₈H₁₅NO₃
MDL Number: MFCD02859909
CAS Number: 57864-64-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR