4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride

Supplier: Matrix Scientific

077834-1G
102667-978EA 400 USD
102667-978
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride
4-Methyl-1,2,3,6-tetrahydropyridine hydrochloride

Hazard: Irritant

Formula: C₆H₁₂ClN
Melting Pt: 145…148 °C
MDL Number: MFCD22123337
CAS Number: 95019-16-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR