5-(Dimethoxymethyl)-2-fluoro-N'-hydroxybenzenecarboximidamide

Supplier: Matrix Scientific

077824-1G
102667-928EA 323.08 USD
102667-928
5-(Dimethoxymethyl)-2-fluoro-N'-hydroxybenzenecarboximidamide
5-(Dimethoxymethyl)-2-fluoro-N'-hydroxybenzenecarboximidamide

Hazard: Irritant

Formula: C₁₀H₁₃FN₂O₃
MDL Number: MFCD22056556
CAS Number: 1365988-79-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR